Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047524.hg.1 |
TC01003052.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047524.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047527.hg.1 |
TC01003052.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047527.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047525.hg.1 |
TC01003052.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047525.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047526.hg.1 |
TC01003052.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047526.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047528.hg.1 |
TC01003052.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047528.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047523.hg.1 |
TC01003052.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047523.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015478.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015478.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015478.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015478.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015478.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015478.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015478.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015478.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015478.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015478.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015478.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015478.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015478.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015478.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015478.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015478.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015478.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015478.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015478.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015478.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215580.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215580.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215580.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215580.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215580.hg.1 |
TC0100015478.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100015478.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015478.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015478.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015478.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015478.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015478.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015478.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015478.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015478.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015478.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015478.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015478.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015478.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015478.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015478.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015478.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015478.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015478.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015478.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015478.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215573.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215573.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215573.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215573.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215573.hg.1 |
TC0100015478.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100215575.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215575.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215575.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215575.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215575.hg.1 |
TC0100015478.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100215576.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215576.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215576.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215576.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215576.hg.1 |
TC0100015478.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100215578.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215578.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215578.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215578.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215578.hg.1 |
TC0100015478.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100215579.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215579.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215579.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215579.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215579.hg.1 |
TC0100015478.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100215581.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215581.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215581.hg.1 |
TC0100015478.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100215582.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215582.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215582.hg.1 |
TC0100015478.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100215577.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215577.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215577.hg.1 |
ENST00000445758.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215577.hg.1 |
NM_001047980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215577.hg.1 |
TC0100015478.hg.1 |
Clariom D TC |