Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010028.hg.1 |
ENST00000409159.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010028.hg.1 |
ENST00000409159.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010028.hg.1 |
ENST00000409159.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
ENST00000311481.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
ENST00000311481.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
ENST00000311481.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
ENST00000311481.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
ENST00000311481.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
ENST00000311481.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
ENST00000311481.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
ENST00000311481.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
ENST00000311481.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
ENST00000311481.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
ENST00000311481.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
ENST00000527397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
ENST00000527397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
ENST00000527397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
ENST00000527397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
ENST00000527397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
ENST00000527397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
ENST00000527397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
ENST00000527397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
ENST00000615245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
ENST00000615245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
ENST00000615245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
ENST00000615245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
ENST00000615245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
ENST00000615245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
ENST00000615245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
ENST00000615245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
ENST00000615245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
ENST00000615245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
NM_030981.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
NM_030981.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
NM_030981.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
NM_030981.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
NM_030981.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
NM_030981.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
NM_030981.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
NM_030981.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
NM_030981.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008067.hg.1 |
NM_030981.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010028.hg.1 |
ENST00000409159.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010028.hg.1 |
ENST00000409159.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010028.hg.1 |
ENST00000409159.2 |
Ensembl |