Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REXO1L1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF495523.1 | - | 1 | 407 | 133 |
Gene Symbol: LOC107986879 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017014126.1 | XP_016869615.1 | 1 | 851 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26552886 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06269788 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06269779 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16113139 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26552883 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26552860 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06269808 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26552874 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06269825 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06269816 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26552868 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26552891 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26552880 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06269822 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06269805 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26552875 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06269782 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06269800 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26552866 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06269818 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26552873 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06269815 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06269827 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26552859 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06269813 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26552871 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06269826 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06269814 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06269821 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26552901 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06269786 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06269809 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06269792 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06269780 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26552876 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26552877 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26552863 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26552898 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06269802 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06269799 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 926151 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 786157 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 461231 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 765112 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 361617 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 98764 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 410948 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 196965 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 384257 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 401491 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 104120 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 253790 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 38338 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 173737 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3258557 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3236267 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3317412 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3339915 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P428373 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3254012 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3224249 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P428373 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 12149 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 58277 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 25982 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 40045 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 4821 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 24445 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 2156 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 12803 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3224249 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3236267 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3254012 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3317412 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P428373 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3339915 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3258557 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3254012 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P428373 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3258557 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3224249 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3317412 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3236267 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3339915 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P428373 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P06269778 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16112699 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16112700 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P428373 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P417713 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_172239_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8151614 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8151611 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8151620 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8151610 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8151604 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8151622 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8151608 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8151618 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8151612 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8151624 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8151626 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8151606 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8151632 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8151605 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8151621 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8151617 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8151613 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8151625 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8151609 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1553009_PM_s_at | - | - |
Affymetrix Human Genome U219 Array | 11740947_at | - | - |
Affymetrix Human Genome U219 Array | 11740947_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11740948_s_at | - | - |
Affymetrix Human Genome U219 Array | 11740948_PM_s_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0800011010.hg.1 | ENST00000603433.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011010.hg.1 | ENST00000603433.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011010.hg.1 | ENST00000619970.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011010.hg.1 | ENST00000619970.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011010.hg.1 | NR_003594.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011010.hg.1 | NR_003594.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011000.hg.1 | ENST00000379010.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011000.hg.1 | ENST00000379010.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011000.hg.1 | ENST00000379010.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011000.hg.1 | ENST00000379010.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011000.hg.1 | ENST00000379010.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011000.hg.1 | ENST00000379010.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011000.hg.1 | ENST00000379010.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011000.hg.1 | ENST00000608646.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011000.hg.1 | ENST00000608646.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011000.hg.1 | ENST00000608646.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011000.hg.1 | ENST00000622202.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011000.hg.1 | ENST00000622202.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011007.hg.1 | ENST00000605109.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011007.hg.1 | ENST00000605109.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011007.hg.1 | ENST00000605109.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011005.hg.1 | ENST00000425429.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011005.hg.1 | ENST00000425429.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011005.hg.1 | ENST00000425429.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011006.hg.1 | ENST00000540724.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011006.hg.1 | ENST00000540724.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011006.hg.1 | ENST00000540724.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011003.hg.1 | ENST00000604264.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011003.hg.1 | ENST00000604264.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011003.hg.1 | ENST00000604264.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011003.hg.1 | ENST00000604264.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011003.hg.1 | ENST00000604264.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011003.hg.1 | ENST00000604264.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011007.hg.1 | ENST00000605109.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011007.hg.1 | ENST00000605109.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011007.hg.1 | ENST00000605109.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011005.hg.1 | ENST00000425429.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011005.hg.1 | ENST00000425429.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011005.hg.1 | ENST00000425429.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011006.hg.1 | ENST00000540724.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011006.hg.1 | ENST00000540724.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011006.hg.1 | ENST00000540724.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011000.hg.1 | ENST00000379010.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011000.hg.1 | ENST00000379010.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011000.hg.1 | ENST00000379010.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011000.hg.1 | ENST00000379010.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011000.hg.1 | ENST00000379010.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011000.hg.1 | ENST00000379010.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011000.hg.1 | ENST00000379010.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011000.hg.1 | ENST00000608646.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011000.hg.1 | ENST00000608646.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011000.hg.1 | ENST00000608646.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011000.hg.1 | ENST00000622202.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011000.hg.1 | ENST00000622202.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011010.hg.1 | ENST00000603433.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011010.hg.1 | ENST00000603433.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011010.hg.1 | ENST00000619970.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011010.hg.1 | ENST00000619970.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011010.hg.1 | NR_003594.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011010.hg.1 | NR_003594.1 | Refseq |
Human Genome U133 Plus 2.0 Array | 1553009_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 1553009_PM_s_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553009_s_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553009_s_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553009_s_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553009_s_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553009_s_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553009_s_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553009_s_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553009_s_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553009_s_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553009_s_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553009_s_at10 | - | - |
Human Genome U95 Set | 63072_at | - | - |
Human Genome U95 Set | 63072_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A035923 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A035922 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A036736 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A035922 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A036736 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A035923 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 710818 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1810639 | NM_172239.4 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1810639 | AF495523.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1810639 | NM_172239.4 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1810639 | AF495523.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1810639 | NM_172239.4 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1810639 | AF495523.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1810639 | NM_172239.4 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1810639 | AF495523.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1810639 | NM_172239.4 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1810639 | AF495523.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_26665874-S | NM_172239.4 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_26665874-S | AF495523.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_5034-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0002850717 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0002850717 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1810639 | NM_172239.4 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1810639 | AF495523.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1810639 | NM_172239.4 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1810639 | AF495523.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 6480474 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_5034 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1810639 | NM_172239.4 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1810639 | AF495523.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1810639 | NM_172239.4 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1810639 | AF495523.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1810639 | NM_172239.4 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1810639 | AF495523.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1810639 | NM_172239.4 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1810639 | AF495523.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1810639 | NM_172239.4 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1810639 | AF495523.1 | human_transcriptome |