Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008390.hg.1 |
ENST00000447693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012819.hg.1 |
ENST00000526184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
ENST00000262306.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
ENST00000262306.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
ENST00000262306.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
ENST00000262306.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
ENST00000262306.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
ENST00000409477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
ENST00000409477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
ENST00000409477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
ENST00000409477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
ENST00000409477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
ENST00000409477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
ENST00000409477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
ENST00000409477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
ENST00000409906.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
ENST00000409906.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
ENST00000409906.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
ENST00000409906.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
ENST00000409906.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
ENST00000409906.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
ENST00000409906.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
ENST00000494946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
ENST00000494946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
ENST00000494946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
ENST00000494946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
ENST00000494946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
ENST00000572954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
ENST00000572954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
ENST00000572954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
NM_007108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
NM_007108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
NM_007108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
NM_007108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
NM_007108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
NM_007108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
NM_007108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
NM_207013.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
NM_207013.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
NM_207013.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
NM_207013.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009137.hg.1 |
NM_207013.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011605.hg.1 |
NM_007108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011605.hg.1 |
NM_207013.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011605.hg.1 |
TC16000791.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011606.hg.1 |
ENST00000262306.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011606.hg.1 |
NM_007108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011606.hg.1 |
NM_207013.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011606.hg.1 |
TC16000791.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1300008390.hg.1 |
ENST00000447693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600169585.hg.1 |
ENST00000262306.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600169585.hg.1 |
ENST00000409477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600169585.hg.1 |
ENST00000409906.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600169585.hg.1 |
ENST00000494946.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600169585.hg.1 |
NM_007108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600169585.hg.1 |
NM_207013.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600169585.hg.1 |
ENST00000262306.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600169585.hg.1 |
ENST00000409477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600169585.hg.1 |
ENST00000409906.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600169585.hg.1 |
ENST00000494946.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600169585.hg.1 |
NM_007108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600169585.hg.1 |
NM_207013.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600169585.hg.1 |
TC1600009137.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600169587.hg.1 |
ENST00000262306.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600169587.hg.1 |
ENST00000409477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600169587.hg.1 |
ENST00000409906.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600169587.hg.1 |
ENST00000494946.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600169587.hg.1 |
ENST00000572954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600169587.hg.1 |
NM_007108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600169587.hg.1 |
NM_207013.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600169587.hg.1 |
ENST00000262306.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600169587.hg.1 |
ENST00000409477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600169587.hg.1 |
ENST00000409906.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600169587.hg.1 |
ENST00000494946.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600169587.hg.1 |
ENST00000572954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600169587.hg.1 |
NM_007108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600169587.hg.1 |
NM_207013.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600169587.hg.1 |
TC1600009137.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100012819.hg.1 |
ENST00000526184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
ENST00000262306.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
ENST00000262306.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
ENST00000262306.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
ENST00000262306.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
ENST00000262306.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
ENST00000409477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
ENST00000409477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
ENST00000409477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
ENST00000409477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
ENST00000409477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
ENST00000409477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
ENST00000409477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
ENST00000409477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
ENST00000409906.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
ENST00000409906.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
ENST00000409906.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
ENST00000409906.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
ENST00000409906.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
ENST00000409906.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
ENST00000409906.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
ENST00000494946.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
ENST00000494946.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
ENST00000494946.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
ENST00000494946.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
ENST00000494946.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
ENST00000572954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
ENST00000572954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
ENST00000572954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
NM_007108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
NM_007108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
NM_007108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
NM_007108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
NM_007108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
NM_007108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
NM_007108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
NM_207013.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
NM_207013.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
NM_207013.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
NM_207013.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009137.hg.1 |
NM_207013.2 |
Refseq |