Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR6V1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001667.1 | NP_001001667.1 | 1 | 850 | 90 | |
GenBank mRNA | BC137044.1 | - | 1 | 871 | 9021040 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon |