Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03013606.hg.1 |
TC03000747.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03013606.hg.1 |
ENST00000383165.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03013606.hg.1 |
NM_001040061.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03013609.hg.1 |
TC03000747.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03013609.hg.1 |
ENST00000383165.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03013609.hg.1 |
NM_001040061.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008962.hg.1 |
ENST00000383165.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008962.hg.1 |
ENST00000383165.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008962.hg.1 |
ENST00000383165.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008962.hg.1 |
ENST00000383165.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008962.hg.1 |
ENST00000383165.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008962.hg.1 |
ENST00000383165.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008962.hg.1 |
ENST00000383165.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008962.hg.1 |
ENST00000470680.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008962.hg.1 |
ENST00000470680.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008962.hg.1 |
ENST00000470680.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008962.hg.1 |
ENST00000470680.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008962.hg.1 |
ENST00000470680.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008962.hg.1 |
ENST00000470680.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008962.hg.1 |
ENST00000498709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008962.hg.1 |
ENST00000498709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008962.hg.1 |
ENST00000498709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008962.hg.1 |
ENST00000498709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008962.hg.1 |
ENST00000498709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008962.hg.1 |
NM_001040061.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008962.hg.1 |
NM_001040061.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008962.hg.1 |
NM_001040061.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008962.hg.1 |
NM_001040061.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008962.hg.1 |
NM_001040061.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008962.hg.1 |
NM_001040061.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008962.hg.1 |
NM_001040061.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008962.hg.1 |
NM_001040061.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164173.hg.1 |
ENST00000383165.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164173.hg.1 |
ENST00000470680.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164173.hg.1 |
ENST00000498709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164173.hg.1 |
NM_001040061.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164173.hg.1 |
ENST00000383165.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164173.hg.1 |
ENST00000470680.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164173.hg.1 |
ENST00000498709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164173.hg.1 |
NM_001040061.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164173.hg.1 |
TC0300008962.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300008962.hg.1 |
ENST00000383165.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008962.hg.1 |
ENST00000383165.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008962.hg.1 |
ENST00000383165.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008962.hg.1 |
ENST00000383165.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008962.hg.1 |
ENST00000383165.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008962.hg.1 |
ENST00000383165.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008962.hg.1 |
ENST00000383165.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008962.hg.1 |
ENST00000470680.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008962.hg.1 |
ENST00000470680.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008962.hg.1 |
ENST00000470680.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008962.hg.1 |
ENST00000470680.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008962.hg.1 |
ENST00000470680.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008962.hg.1 |
ENST00000470680.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008962.hg.1 |
ENST00000498709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008962.hg.1 |
ENST00000498709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008962.hg.1 |
ENST00000498709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008962.hg.1 |
ENST00000498709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008962.hg.1 |
ENST00000498709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008962.hg.1 |
NM_001040061.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008962.hg.1 |
NM_001040061.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008962.hg.1 |
NM_001040061.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008962.hg.1 |
NM_001040061.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008962.hg.1 |
NM_001040061.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008962.hg.1 |
NM_001040061.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008962.hg.1 |
NM_001040061.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008962.hg.1 |
NM_001040061.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164170.hg.1 |
ENST00000383165.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164170.hg.1 |
ENST00000470680.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164170.hg.1 |
NM_001040061.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164170.hg.1 |
ENST00000383165.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164170.hg.1 |
ENST00000470680.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164170.hg.1 |
NM_001040061.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164170.hg.1 |
TC0300008962.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300164169.hg.1 |
ENST00000383165.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164169.hg.1 |
ENST00000470680.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164169.hg.1 |
NM_001040061.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164169.hg.1 |
ENST00000383165.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164169.hg.1 |
ENST00000470680.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164169.hg.1 |
NM_001040061.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164169.hg.1 |
TC0300008962.hg.1 |
Clariom D TC |