Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
small nucleolar RNA, C/D box 14E |
SNORD14E | -479.0 | Sense | Chr. 11 - 123058077 - 123058156 on NCBI Build GRCh38 | |
Downstream |
small nucleolar RNA, C/D box 14C |
SNORD14C | 73.0 | Sense | Chr. 11 - 123059335 - 123059422 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |