Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038443.1 | 1 | 1752 | 146 | ||
GenBank mRNA | BC066922.1 | - | 1 | 1752 | 146 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | sarcoglycan delta | SGCD |
35DAG, CMD1L, DAGD, SG-delta, SGCDP, SGD |
-82.8kb | Sense | Chr. 5 - 155728636 - 156767788 on NCBI Build GRCh38 |
Downstream | T-cell immunoglobulin and mucin domain containing 4 | TIMD4 |
SMUCKLER, TIM4 |
66.8kb | Anti-sense | Chr. 5 - 156915750 - 156963255 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04395034 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24574481 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24574433 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24574486 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24574487 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24574494 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04395042 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24574495 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04395037 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24574455 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04395016 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24574447 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04395046 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24574435 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04395050 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24574496 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24574432 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24574489 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15755481 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24574434 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24574443 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04395029 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24574490 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04395024 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15755478 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04395022 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24574500 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24574431 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15755480 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24574430 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04395057 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24574502 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15755526 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15755521 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24574493 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04395056 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24574484 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04395028 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04395023 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 169012 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3231771 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3719 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 43635 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3215720 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3306964 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3231771 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3231771 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04395026 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15755499 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04395029 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15755506 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04395034 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04395036 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04395022 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04395031 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24574436 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04395030 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24574462 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15755478 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24574447 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24574455 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24574446 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04395024 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15755481 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04395035 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15755503 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15755503 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04395031 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15755481 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24574462 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24574447 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04395022 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04395026 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24574446 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15755506 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24574455 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04395029 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04395030 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15755478 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04395036 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15755499 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04395035 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04395024 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04395034 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24574436 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04395021 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04395016 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04395022 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15755481 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04395025 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04395059 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15755520 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15755522 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04395040 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04395014 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04395054 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04395058 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04395046 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04395035 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04395034 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04395050 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15755523 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04395053 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04395029 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04395056 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04395031 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04395019 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04395026 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04395038 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15755525 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NR_002168_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8109506 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8109505 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A029076 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A029076 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 158516 | - | - |
ABI Human Genome Survey Microarray Version 2.0 | 234194 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_2045610 | BC066922.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2045610 | NR_038443.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2045610 | BC066922.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2045610 | NR_038443.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2045610 | BC066922.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2045610 | NR_038443.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2045610 | BC066922.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2045610 | NR_038443.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2045610 | BC066922.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2045610 | NR_038443.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2045610 | BC066922.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2045610 | NR_038443.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2045610 | BC066922.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2045610 | NR_038443.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2045610 | BC066922.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2045610 | NR_038443.1 | human_transcriptome |