Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000274820.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000274820.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000412023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000412023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000412023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000412023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000412023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000412023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000412023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000412023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000412023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000421824.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000421824.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000421824.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000421824.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000421824.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000421824.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000421824.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000441655.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000441655.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000441655.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000451612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000451612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000451612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000451612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000504533.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000504533.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000602547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000602547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000602547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000602547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000602547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
ENST00000602547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
NR_002803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
NR_002803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
NR_002803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
NR_033796.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
NR_033796.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
NR_033796.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
NR_033796.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
NR_033796.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012589.hg.1 |
NR_033796.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001215.hg.1 |
ENST00000412023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001215.hg.1 |
NR_002803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001215.hg.1 |
TC12000097.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200198995.hg.1 |
ENST00000412023.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200198995.hg.1 |
ENST00000441655.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200198995.hg.1 |
NR_002803.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200198995.hg.1 |
ENST00000412023.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200198995.hg.1 |
ENST00000441655.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200198995.hg.1 |
NR_002803.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200198995.hg.1 |
TC1200012589.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000274820.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000274820.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000412023.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000412023.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000412023.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000412023.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000412023.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000412023.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000412023.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000412023.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000412023.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000421824.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000421824.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000421824.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000421824.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000421824.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000421824.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000421824.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000441655.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000441655.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000441655.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000451612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000451612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000451612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000451612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000504533.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000504533.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000602547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000602547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000602547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000602547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000602547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
ENST00000602547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
NR_002803.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
NR_002803.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
NR_002803.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
NR_033796.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
NR_033796.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
NR_033796.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
NR_033796.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
NR_033796.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012589.hg.1 |
NR_033796.2 |
Refseq |