Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR2T35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001827.1 | NP_001001827.1 | 1 | 353 | 128 |
Gene Symbol: OR2T2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004136.1 | NP_001004136.1 | 1 | 356 | 131 | |
GenBank mRNA | BC136961.1 | - | 1 | 378 | 9020953 | 131 |
BC136962.1 | - | 1 | 378 | 9020954 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21007736 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006900 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15206577 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15206578 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15205980 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15205853 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15206571 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00932250 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006904 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21007002 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15206591 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006958 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 362575 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 400777 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 276278 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3336014 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0294 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0295 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0296 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 13683 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3336014 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3336014 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3336014 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15206587 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006904 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006958 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006892 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15206585 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15206563 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006893 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15205883 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006911 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15206564 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15206511 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15206577 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21007729 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006900 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15205873 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15206571 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15206578 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15206583 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006896 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006904 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006896 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15206587 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15206577 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15206571 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15206583 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006911 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15205873 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15206564 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15206585 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006893 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006900 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15206578 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006892 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15205883 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15206563 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21007729 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15206511 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006958 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00932245 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00932246 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00932523 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P35545117 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00932509 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00932247 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00932507 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00932506 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00932248 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00932502 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00932249 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00932523 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00932512 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00932499 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00932513 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00932496 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00932245 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 7911282 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7911282 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7925758 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7925758 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7911281 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7911281 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7925757 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7925757 | - | - |
Affymetrix Human Genome U219 Array | 11759278_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11759278_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0100012358.hg.1 | ENST00000342927.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012358.hg.1 | NM_001004136.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018134.hg.1 | ENST00000317450.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018134.hg.1 | NM_001001827.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01031135.hg.1 | TC01002038.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01031135.hg.1 | ENST00000342927.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01031135.hg.1 | NM_001004136.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01061507.hg.1 | TC01004090.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01061507.hg.1 | ENST00000317450.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01061507.hg.1 | NM_001001827.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012358.hg.1 | ENST00000342927.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012358.hg.1 | NM_001004136.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018134.hg.1 | ENST00000317450.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018134.hg.1 | NM_001001827.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100236349.hg.1 | ENST00000317450.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100236349.hg.1 | NM_001001827.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100236349.hg.1 | ENST00000317450.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100236349.hg.1 | NM_001001827.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100236349.hg.1 | TC0100018134.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100190637.hg.1 | ENST00000342927.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190637.hg.1 | NM_001004136.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190637.hg.1 | ENST00000342927.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190637.hg.1 | NM_001004136.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190637.hg.1 | TC0100012358.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100190638.hg.1 | TC0100012358.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A013779 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A013779 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A013779 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A013779 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1671386 | - | - |
HumanHT-12 v4 Expression BeadChip | ILMN_2148122 | NM_001001827.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1777332 | NM_001001827.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1777332 | NM_001004136.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1777332 | BC136961.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1777332 | BC136962.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1795254 | NM_001001827.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1795254 | NM_001004136.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1795254 | BC136961.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1795254 | BC136962.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1671386 | - | - |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1777332 | NM_001001827.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1777332 | NM_001004136.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1777332 | BC136961.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1777332 | BC136962.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1700439 | - | - |
HumanHT-12 V4.0 expression beadchip | ILMN_2148122 | NM_001001827.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1795254 | NM_001001827.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1795254 | NM_001004136.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1795254 | BC136961.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1795254 | BC136962.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2148122 | NM_001001827.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2148122 | NM_001001827.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_16988-S | - | - |
Illumina HumanRef-8 v2 Expression BeadChip | GI_18689-A | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1777332 | NM_001001827.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1777332 | NM_001004136.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1777332 | BC136961.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1777332 | BC136962.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1671386 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1700439 | - | - |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1700439 | - | - |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1777332 | NM_001001827.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1777332 | NM_001004136.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1777332 | BC136961.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1777332 | BC136962.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 7380246 | - | - |
Illumina Sentrix Human-6 v2 Expression BeadChip | 520315 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_18689 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_16988 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2148122 | NM_001001827.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1671386 | - | - |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2148122 | NM_001001827.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1777332 | NM_001001827.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1777332 | NM_001004136.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1777332 | BC136961.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1777332 | BC136962.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1795254 | NM_001001827.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1795254 | NM_001004136.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1795254 | BC136961.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1795254 | BC136962.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1671386 | - | - |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2148122 | NM_001001827.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1777332 | NM_001001827.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1777332 | NM_001004136.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1777332 | BC136961.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1777332 | BC136962.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1795254 | NM_001001827.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1795254 | NM_001004136.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1795254 | BC136961.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1795254 | BC136962.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1700439 | - | - |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2148122 | NM_001001827.1 | human_transcriptome |