Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000605881.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000605881.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000605881.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000605881.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000605881.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000605881.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000605881.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000605881.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000605881.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000605881.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000605881.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000605881.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000605881.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000605881.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |