Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000526541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000526541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000526541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000526541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000526541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000526541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000526541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000526541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000526541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
ENST00000526541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
NR_002720.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
NR_002720.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
NR_002720.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
NR_002720.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
NR_002720.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
NR_002720.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
NR_002720.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
NR_002720.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
NR_002720.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006628.hg.1 |
NR_002720.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000451043.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000526541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000526541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000526541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000526541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000526541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000526541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000526541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000526541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000526541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
ENST00000526541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
NR_002720.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
NR_002720.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
NR_002720.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
NR_002720.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
NR_002720.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
NR_002720.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
NR_002720.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
NR_002720.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
NR_002720.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006628.hg.1 |
NR_002720.2 |
Refseq |