Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNQ1OT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_002728.3 | 1 | 81 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | transient receptor potential cation channel subfamily M member 5 | TRPM5 |
LTRPC5, MTR1 |
-247.9kb | Anti-sense | Chr. 11 - 2403962 - 2423424 on NCBI Build GRCh38 |
Downstream | KCNQ1 antisense RNA 1 | KCNQ1-AS1 | 156.8kb | Anti-sense | Chr. 11 - 2840135 - 2861569 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |