Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_002216.1 | 11 - 12 | 1362 | 83 | ||
GenBank mRNA | AK124700.1 | - | 11 - 12 | 1362 | 83 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | thymosin beta 15a | TMSB15A |
TMSB15, TMSB15B, TMSL8, TMSNB, Tb15, TbNB |
-49.4kb | Anti-sense | Chr. X - 102513682 - 102516771 on NCBI Build GRCh38 |
Downstream |
armadillo repeat containing, X-linked 5 |
ARMCX5 | GASP5 | 32.8kb | Sense | Chr. X - 102599168 - 102604159 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons |