Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021724.hg.1 |
TC07001395.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021724.hg.1 |
ENST00000352861.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021724.hg.1 |
ENST00000415949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021724.hg.1 |
NM_014302.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015285.hg.1 |
ENST00000448883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021725.hg.1 |
TC07001395.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021725.hg.1 |
ENST00000352861.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021725.hg.1 |
ENST00000415949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021725.hg.1 |
NM_014302.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021714.hg.1 |
TC07001395.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021714.hg.1 |
ENST00000352861.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021714.hg.1 |
ENST00000395535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021714.hg.1 |
ENST00000415949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021714.hg.1 |
ENST00000450622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021714.hg.1 |
NM_001012456.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021714.hg.1 |
NM_014302.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021726.hg.1 |
TC07001395.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021726.hg.1 |
ENST00000352861.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021726.hg.1 |
ENST00000395535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021726.hg.1 |
ENST00000415949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021726.hg.1 |
NM_001012456.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021726.hg.1 |
NM_014302.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
ENST00000352861.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
ENST00000352861.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
ENST00000352861.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
ENST00000395535.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
ENST00000395535.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
ENST00000395535.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
ENST00000415949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
ENST00000415949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
ENST00000415949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
ENST00000415949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
ENST00000415949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
ENST00000450622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
ENST00000450622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
ENST00000450622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
ENST00000450622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
ENST00000460484.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
ENST00000460484.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
ENST00000460484.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
ENST00000460484.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
ENST00000460484.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
ENST00000461536.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
ENST00000461536.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
ENST00000480303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
ENST00000480303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
ENST00000480303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
ENST00000480303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
NM_001012456.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
NM_001012456.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
NM_001012456.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
NM_014302.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
NM_014302.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011119.hg.1 |
NM_014302.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700179526.hg.1 |
ENST00000352861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700179526.hg.1 |
ENST00000395535.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700179526.hg.1 |
ENST00000415949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700179526.hg.1 |
ENST00000450622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700179526.hg.1 |
ENST00000460484.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700179526.hg.1 |
ENST00000480303.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700179526.hg.1 |
NM_001012456.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700179526.hg.1 |
NM_014302.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700179526.hg.1 |
ENST00000352861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700179526.hg.1 |
ENST00000395535.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700179526.hg.1 |
ENST00000415949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700179526.hg.1 |
ENST00000450622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700179526.hg.1 |
ENST00000460484.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700179526.hg.1 |
ENST00000480303.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700179526.hg.1 |
NM_001012456.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700179526.hg.1 |
NM_014302.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700179526.hg.1 |
TC0700011119.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200015285.hg.1 |
ENST00000448883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
ENST00000352861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
ENST00000352861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
ENST00000352861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
ENST00000395535.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
ENST00000395535.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
ENST00000395535.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
ENST00000415949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
ENST00000415949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
ENST00000415949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
ENST00000415949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
ENST00000415949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
ENST00000450622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
ENST00000450622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
ENST00000450622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
ENST00000450622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
ENST00000460484.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
ENST00000460484.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
ENST00000460484.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
ENST00000460484.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
ENST00000460484.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
ENST00000461536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
ENST00000461536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
ENST00000480303.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
ENST00000480303.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
ENST00000480303.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
ENST00000480303.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
NM_001012456.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
NM_001012456.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
NM_001012456.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
NM_014302.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
NM_014302.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011119.hg.1 |
NM_014302.3 |
Refseq |