Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024723.hg.1 |
ENST00000432826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024723.hg.1 |
ENST00000446034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024723.hg.1 |
NM_001135686.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024723.hg.1 |
TC19001830.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024724.hg.1 |
ENST00000432826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024724.hg.1 |
ENST00000446034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024724.hg.1 |
NM_001135686.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024724.hg.1 |
TC19001830.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011390.hg.1 |
ENST00000432826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011390.hg.1 |
ENST00000432826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011390.hg.1 |
ENST00000432826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011390.hg.1 |
ENST00000432826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011390.hg.1 |
ENST00000432826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011390.hg.1 |
ENST00000432826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011390.hg.1 |
ENST00000432826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011390.hg.1 |
ENST00000432826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011390.hg.1 |
ENST00000616041.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011390.hg.1 |
ENST00000616041.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011390.hg.1 |
ENST00000616041.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011390.hg.1 |
ENST00000616041.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011390.hg.1 |
ENST00000616041.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011390.hg.1 |
ENST00000616041.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011390.hg.1 |
ENST00000616041.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011390.hg.1 |
ENST00000616041.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024725.hg.1 |
ENST00000413865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024725.hg.1 |
ENST00000432826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024725.hg.1 |
ENST00000446034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024725.hg.1 |
NM_001135686.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024725.hg.1 |
TC19001830.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024729.hg.1 |
ENST00000432826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024729.hg.1 |
NM_001135686.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024729.hg.1 |
TC19001830.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024726.hg.1 |
ENST00000413865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024726.hg.1 |
ENST00000432826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024726.hg.1 |
ENST00000446034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024726.hg.1 |
NM_001135686.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024726.hg.1 |
TC19001830.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1900011390.hg.1 |
ENST00000432826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011390.hg.1 |
ENST00000432826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011390.hg.1 |
ENST00000432826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011390.hg.1 |
ENST00000432826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011390.hg.1 |
ENST00000432826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011390.hg.1 |
ENST00000432826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011390.hg.1 |
ENST00000432826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011390.hg.1 |
ENST00000432826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011390.hg.1 |
ENST00000616041.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011390.hg.1 |
ENST00000616041.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011390.hg.1 |
ENST00000616041.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011390.hg.1 |
ENST00000616041.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011390.hg.1 |
ENST00000616041.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011390.hg.1 |
ENST00000616041.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011390.hg.1 |
ENST00000616041.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011390.hg.1 |
ENST00000616041.4 |
Ensembl |