Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001335.hg.1 |
TC03000058.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001335.hg.1 |
ENST00000439539.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001335.hg.1 |
NR_004431.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001335.hg.1 |
NR_024144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001335.hg.1 |
NR_024145.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001336.hg.1 |
TC03000058.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001336.hg.1 |
NR_004431.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001336.hg.1 |
NR_024144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001336.hg.1 |
NR_024145.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145723.hg.1 |
ENST00000439539.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145723.hg.1 |
ENST00000605014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145723.hg.1 |
ENST00000605014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145723.hg.1 |
NR_004431.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145723.hg.1 |
NR_024144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145723.hg.1 |
NR_024145.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145723.hg.1 |
NR_073566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145723.hg.1 |
NR_073567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145723.hg.1 |
ENST00000439539.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145723.hg.1 |
ENST00000605014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145723.hg.1 |
ENST00000605014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145723.hg.1 |
NR_004431.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145723.hg.1 |
NR_024144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145723.hg.1 |
NR_024145.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145723.hg.1 |
NR_073566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145723.hg.1 |
NR_073567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145723.hg.1 |
TC0300006582.hg.1 |
Clariom D TC |