Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACBD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301251.1 | NP_001288180.1 | 2 - 3 | 248 | 77 | |
NM_001301252.1 | NP_001288181.1 | 1 - 2 | 562 | 77 | ||
NM_001301253.1 | NP_001288182.1 | 2 - 3 | 238 | 77 | ||
NM_001301254.1 | NP_001288183.1 | 1 - 2 | 562 | 77 | ||
XM_017016900.1 | XP_016872389.1 | 2 - 3 | 278 | 77 | ||
XM_017016901.1 | XP_016872390.1 | 2 - 3 | 258 | 77 | ||
XM_017016902.1 | XP_016872391.1 | 2 - 3 | 277 | 77 | ||
XM_017016903.1 | XP_016872392.1 | 2 - 3 | 262 | 77 | ||
XM_017016904.1 | XP_016872393.1 | 2 - 3 | 290 | 77 | ||
XM_017016905.1 | XP_016872394.1 | 2 - 3 | 276 | 77 | ||
XM_017016907.1 | XP_016872396.1 | 2 - 3 | 290 | 77 | ||
XM_017016908.1 | XP_016872397.1 | 2 - 3 | 279 | 77 | ||
GenBank mRNA | AB082527.1 | - | 2 - 3 | 197 | 77 | |
AF505653.1 | - | 2 - 3 | 235 | 77 | ||
AK294510.1 | - | 1 - 2 | 480 | 77 | ||
AK304842.1 | - | 1 - 2 | 213 | 77 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microtubule associated serine/threonine kinase like | MASTL |
GREATWALL, GW, GWL, MAST-L, THC2 |
-44.8kb | Anti-sense | Chr. 10 - 27154468 - 27187953 on NCBI Build GRCh38 |
Downstream |
leucine rich repeat containing 37 member A6, pseudogene |
LRRC37A6P | LRRC37E | 5.4kb | Sense | Chr. 10 - 27245853 - 27252306 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Probe spans exons |