Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | calcium regulated heat stable protein 1 | CARHSP1 |
CRHSP-24, CRHSP24, CSDC1 |
Sense | Chr. 16 - 8852942 - 8869012 on NCBI Build GRCh38 | |
Downstream | lipopolysaccharide-induced tumor necrosis factor-alpha factor-like | LOC101929989 | 17.0kb | Anti-sense | Chr. 16 - 8881975 - 8892284 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |