Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000465768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000465768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000465768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000493621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000493621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000493621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000494795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000494795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000494795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000494795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000494795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000494795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000465768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000465768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000465768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000493621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000493621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000493621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000494795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000494795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000494795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000494795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000494795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000494795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |