Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY358685.1 | - | 1 | 252 | 74 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | C-type lectin domain family 1 member A | CLEC1A |
CLEC-1, CLEC1 |
-27.1kb | Sense | Chr. 12 - 10069504 - 10099065 on NCBI Build GRCh38 |
Downstream | oxidized low density lipoprotein receptor 1 | OLR1 |
CLEC8A, LOX1, LOXIN, SCARE1, SLOX1 |
31.5kb | Sense | Chr. 12 - 10158300 - 10172191 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |