Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AL137486.1 | - | 1 | 942 | 73 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | zinc finger protein 470 | ZNF470 | CZF-1 | -35.5kb | Sense | Chr. 19 - 56567521 - 56582894 on NCBI Build GRCh38 |
Downstream | small integral membrane protein 17 | SMIM17 | 16.7kb | Sense | Chr. 19 - 56639251 - 56657247 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |