Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
ENST00000419083.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
ENST00000419083.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
ENST00000419083.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
ENST00000419083.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
ENST00000419083.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
ENST00000419083.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
ENST00000419083.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
ENST00000419083.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
ENST00000419083.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
ENST00000419083.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
ENST00000419083.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
ENST00000419083.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
ENST00000420452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
ENST00000420452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
ENST00000420452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
ENST00000420452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
ENST00000420452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
ENST00000420452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
ENST00000420452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
ENST00000420452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
ENST00000439180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
ENST00000439180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
ENST00000439180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
ENST00000439180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
ENST00000448719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
ENST00000448719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
ENST00000448719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
NR_026766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
NR_026766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
NR_026766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
NR_026766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
NR_026766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
NR_026766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
NR_110230.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
NR_110230.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
NR_110230.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
NR_110230.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016634.hg.1 |
NR_110230.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
ENST00000419083.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
ENST00000419083.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
ENST00000419083.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
ENST00000419083.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
ENST00000419083.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
ENST00000419083.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
ENST00000419083.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
ENST00000419083.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
ENST00000419083.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
ENST00000419083.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
ENST00000419083.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
ENST00000419083.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
ENST00000420452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
ENST00000420452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
ENST00000420452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
ENST00000420452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
ENST00000420452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
ENST00000420452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
ENST00000420452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
ENST00000420452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
ENST00000439180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
ENST00000439180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
ENST00000439180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
ENST00000439180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
ENST00000448719.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
ENST00000448719.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
ENST00000448719.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
NR_026766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
NR_026766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
NR_026766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
NR_026766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
NR_026766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
NR_026766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
NR_110230.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
NR_110230.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
NR_110230.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
NR_110230.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016634.hg.1 |
NR_110230.1 |
Refseq |