Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000219542.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000219542.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000219542.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000219542.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000219542.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000219542.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000219542.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000219542.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000219542.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000564661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000564661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000564661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000564661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000564661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000564661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000564661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000564661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000564661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000564661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000564661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000567076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000567076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000567076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000567076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000567076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000568415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000568415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000568415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000568415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000568415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000568415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000568415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000570132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000570132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000570132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000570132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000570132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000570132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
ENST00000570132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
NM_024042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
NM_024042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
NM_024042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
NM_024042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
NM_024042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
NM_024042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
NM_024042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
NM_024042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
NM_024042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
NM_024042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
NM_024042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
NM_024042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006492.hg.1 |
NM_024042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000219542.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000219542.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000219542.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000219542.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000219542.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000219542.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000219542.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000219542.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000219542.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000564661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000564661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000564661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000564661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000564661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000564661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000564661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000564661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000564661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000564661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000564661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000567076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000567076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000567076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000567076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000567076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000568223.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000568415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000568415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000568415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000568415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000568415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000568415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000568415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000570132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000570132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000570132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000570132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000570132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000570132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
ENST00000570132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
NM_024042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
NM_024042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
NM_024042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
NM_024042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
NM_024042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
NM_024042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
NM_024042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
NM_024042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
NM_024042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
NM_024042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
NM_024042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
NM_024042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006492.hg.1 |
NM_024042.3 |
Refseq |