Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007115.hg.1 |
TC03000332.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007115.hg.1 |
ENST00000305690.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007115.hg.1 |
ENST00000354773.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007115.hg.1 |
ENST00000436784.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007115.hg.1 |
ENST00000461183.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007115.hg.1 |
ENST00000471180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007115.hg.1 |
ENST00000473032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007115.hg.1 |
ENST00000477382.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007115.hg.1 |
NM_001144951.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007115.hg.1 |
NM_145262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007115.hg.1 |
NR_026699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007115.hg.1 |
NR_026700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007115.hg.1 |
NR_026701.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007115.hg.1 |
NR_026702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007105.hg.1 |
TC03000332.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007105.hg.1 |
ENST00000305690.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007105.hg.1 |
ENST00000354773.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007105.hg.1 |
ENST00000473032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007105.hg.1 |
NM_001144951.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007105.hg.1 |
NM_145262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007105.hg.1 |
NR_026699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007105.hg.1 |
NR_026700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007105.hg.1 |
NR_026701.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007105.hg.1 |
NR_026702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007106.hg.1 |
TC03000332.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007106.hg.1 |
ENST00000305690.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007106.hg.1 |
ENST00000354773.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007106.hg.1 |
ENST00000471180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007106.hg.1 |
ENST00000473032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007106.hg.1 |
NM_001144951.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007106.hg.1 |
NM_145262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007106.hg.1 |
NR_026699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007106.hg.1 |
NR_026700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007106.hg.1 |
NR_026701.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007106.hg.1 |
NR_026702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
ENST00000305690.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
ENST00000436784.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
ENST00000461183.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
ENST00000471180.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
ENST00000473032.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
ENST00000473583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
ENST00000477382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
ENST00000486393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
ENST00000489173.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
NM_001144951.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
NM_145262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
NR_026699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
NR_026700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
NR_026701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
NR_026702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
ENST00000305690.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
ENST00000436784.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
ENST00000461183.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
ENST00000471180.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
ENST00000473032.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
ENST00000473583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
ENST00000477382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
ENST00000486393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
ENST00000489173.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
NM_001144951.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
NM_145262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
NR_026699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
NR_026700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
NR_026701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
NR_026702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153711.hg.1 |
TC0300007472.hg.1 |
Clariom D TC |