Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000113.hg.1 |
ENST00000420381.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000113.hg.1 |
ENST00000428780.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000113.hg.1 |
ENST00000434470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000113.hg.1 |
NR_024628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000113.hg.1 |
NR_027708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000113.hg.1 |
NR_027709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000113.hg.1 |
TC10000009.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000144281.hg.1 |
ENST00000420381.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000144281.hg.1 |
ENST00000420381.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000144281.hg.1 |
ENST00000428780.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000144281.hg.1 |
ENST00000434470.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000144281.hg.1 |
NR_024628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000144281.hg.1 |
NR_027708.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000144281.hg.1 |
NR_027709.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000144281.hg.1 |
ENST00000420381.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000144281.hg.1 |
ENST00000420381.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000144281.hg.1 |
ENST00000428780.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000144281.hg.1 |
ENST00000434470.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000144281.hg.1 |
NR_024628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000144281.hg.1 |
NR_027708.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000144281.hg.1 |
NR_027709.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000144281.hg.1 |
TC1000006465.hg.1 |
Clariom D TC |