Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014038.hg.1 |
ENST00000370307.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014038.hg.1 |
ENST00000370313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014038.hg.1 |
ENST00000370316.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014038.hg.1 |
ENST00000466885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014038.hg.1 |
NM_152864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014038.hg.1 |
TC20001027.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370307.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370307.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370307.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370307.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370307.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370307.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370307.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370316.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370316.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370316.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370316.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370316.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370316.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370316.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370316.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370316.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370316.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370316.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370317.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370317.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370317.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370317.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370317.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000370317.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000461738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000461738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000461738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000466885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000466885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000466885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000466885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000466885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000466885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000466885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000470246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000470246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000470246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000470246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000470246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000472670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000472670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000472670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000486495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000486495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000486495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000486495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000486495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
ENST00000486495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
NM_152864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
NM_152864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
NM_152864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
NM_152864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
NM_152864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
NM_152864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
NM_152864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
NM_152864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
NM_152864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
NM_152864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009802.hg.1 |
NM_152864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168347.hg.1 |
ENST00000370307.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168347.hg.1 |
ENST00000370313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168347.hg.1 |
ENST00000370316.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168347.hg.1 |
ENST00000370317.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168347.hg.1 |
ENST00000461738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168347.hg.1 |
ENST00000466885.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168347.hg.1 |
ENST00000486495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168347.hg.1 |
NM_152864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168347.hg.1 |
TC2000009802.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168347.hg.1 |
ENST00000370307.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168347.hg.1 |
ENST00000370313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168347.hg.1 |
ENST00000370316.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168347.hg.1 |
ENST00000370317.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168347.hg.1 |
ENST00000461738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168347.hg.1 |
ENST00000466885.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168347.hg.1 |
ENST00000486495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168347.hg.1 |
NM_152864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370307.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370307.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370307.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370307.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370307.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370307.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370307.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370316.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370316.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370316.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370316.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370316.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370316.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370316.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370316.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370316.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370316.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370316.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370317.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370317.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370317.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370317.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370317.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000370317.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000461738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000461738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000461738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000466885.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000466885.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000466885.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000466885.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000466885.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000466885.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000466885.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000470246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000470246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000470246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000470246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000470246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000472670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000472670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000472670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000486495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000486495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000486495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000486495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000486495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
ENST00000486495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
NM_152864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
NM_152864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
NM_152864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
NM_152864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
NM_152864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
NM_152864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
NM_152864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
NM_152864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
NM_152864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
NM_152864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009802.hg.1 |
NM_152864.3 |
Refseq |