Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | solute carrier organic anion transporter family member 1B3 | SLCO1B3 |
HBLRR, LST-2, LST-3TM13, LST3, OATP-8, OATP1B3, OATP8, SLC21A8 |
-13.4kb | Anti-sense | Chr. 12 - 20810704 - 20916911 on NCBI Build GRCh38 |
Downstream | solute carrier organic anion transporter family member 1B7 (putative) | SLCO1B7 |
LST-3, LST-3TM12, LST3, SLC21A21 |
83.9kb | Anti-sense | Chr. 12 - 21015696 - 21090106 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |