Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | intersectin 2 | ITSN2 |
PRO2015, SH3D1B, SH3P18, SWA, SWAP |
-165.7kb | Sense | Chr. 2 - 24202866 - 24360528 on NCBI Build GRCh38 |
Downstream | peptidyl-tRNA hydrolase domain containing 1 | PTRHD1 | C2orf79 | 263.6kb | Sense | Chr. 2 - 24790267 - 24793382 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |