Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
protein tyrosine phosphatase, non-receptor type 20 |
PTPN20 |
CT126, PTPN20A, PTPN20B, bA142I17.1, bA42B19.1 |
-134.0kb | Sense | Chr. 10 - 46911134 - 47003834 on NCBI Build GRCh38 |
Downstream | growth differentiation factor 10 | GDF10 |
BMP-3b, BMP3B |
132.7kb | Sense | Chr. 10 - 47300224 - 47313577 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |