Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010854.hg.1 |
ENST00000242208.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010854.hg.1 |
ENST00000242208.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010854.hg.1 |
ENST00000242208.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010854.hg.1 |
ENST00000242208.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010854.hg.1 |
ENST00000242208.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010854.hg.1 |
ENST00000242208.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010854.hg.1 |
ENST00000242208.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010854.hg.1 |
ENST00000242208.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010854.hg.1 |
ENST00000242208.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010854.hg.1 |
ENST00000442711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010854.hg.1 |
ENST00000442711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010854.hg.1 |
ENST00000442711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010854.hg.1 |
ENST00000442711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010854.hg.1 |
ENST00000442711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010854.hg.1 |
ENST00000464515.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010854.hg.1 |
ENST00000464515.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010854.hg.1 |
ENST00000464515.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010854.hg.1 |
NM_002192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010854.hg.1 |
NM_002192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010854.hg.1 |
NM_002192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010854.hg.1 |
NM_002192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010854.hg.1 |
NM_002192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010854.hg.1 |
NM_002192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010854.hg.1 |
NM_002192.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010854.hg.1 |
ENST00000242208.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010854.hg.1 |
ENST00000242208.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010854.hg.1 |
ENST00000242208.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010854.hg.1 |
ENST00000242208.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010854.hg.1 |
ENST00000242208.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010854.hg.1 |
ENST00000242208.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010854.hg.1 |
ENST00000242208.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010854.hg.1 |
ENST00000242208.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010854.hg.1 |
ENST00000242208.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010854.hg.1 |
ENST00000442711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010854.hg.1 |
ENST00000442711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010854.hg.1 |
ENST00000442711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010854.hg.1 |
ENST00000442711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010854.hg.1 |
ENST00000442711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010854.hg.1 |
ENST00000464515.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010854.hg.1 |
ENST00000464515.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010854.hg.1 |
ENST00000464515.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010854.hg.1 |
NM_002192.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010854.hg.1 |
NM_002192.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010854.hg.1 |
NM_002192.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010854.hg.1 |
NM_002192.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010854.hg.1 |
NM_002192.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010854.hg.1 |
NM_002192.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010854.hg.1 |
NM_002192.3 |
Refseq |