Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cystatin 8 | CST8 |
CRES, CTES5 |
-9.8kb | Anti-sense | Chr. 20 - 23491077 - 23507124 on NCBI Build GRCh38 |
Downstream |
cystatin 13, pseudogene |
CST13P |
CSTT, CTES6 |
1.1kb | Anti-sense | Chr. 20 - 23519146 - 23542018 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |