Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | SPANX family member B1 | SPANXB1 |
B1, CT11.2, SPANX-B, SPANXB, SPANXB2, SPANXF1, SPANXF2 |
-169.5kb | Anti-sense | Chr. X - 141002591 - 141003706 on NCBI Build GRCh38 |
Downstream | leucine zipper down-regulated in cancer 1 | LDOC1 |
BCUR1, Mar7, Mart7 |
2.2kb | Sense | Chr. X - 141175745 - 141177214 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |