Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | olfactory receptor family 52 subfamily A member 4 pseudogene | OR52A4P | OR52A4 | -1.1kb | Anti-sense | Chr. 11 - 5120664 - 5121578 on NCBI Build GRCh38 |
Downstream | olfactory receptor family 52 subfamily A member 5 | OR52A5 | OR11-33 | 8.2kb | Anti-sense | Chr. 11 - 5131692 - 5132642 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Homo sapiens Probe spans exons |