Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 1551 | LINC01551 |
C14orf23, c14_5148 |
-1.1mb | Anti-sense | Chr. 14 - 28772704 - 28794794 on NCBI Build GRCh38 |
Downstream | G2/M-phase specific E3 ubiquitin protein ligase | G2E3 |
KIAA1333, PHF7B |
694.9kb | Anti-sense | Chr. 14 - 30559123 - 30620064 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |