Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
golgin A8 family member C, pseudogene |
GOLGA8CP | GOLGA8C | -70.2kb | Anti-sense | Chr. 15 - 20562349 - 20575703 on NCBI Build GRCh38 |
Downstream | neurobeachin pseudogene 1 | NBEAP1 |
BCL8, BCL8A |
23.1kb | Sense | Chr. 15 - 20669468 - 20756151 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |