Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | pituitary tumor-transforming gene 1 protein-interacting protein | LOC102723899 | -164.5kb | Sense | Chr. 7 - 90403379 - 90513397 on NCBI Build GRCh38 | |
Downstream | frizzled class receptor 1 | FZD1 | 586.5kb | Sense | Chr. 7 - 91264468 - 91268817 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |