Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | chromosome 3 open reading frame 20 | C3orf20 | Sense | Chr. 3 - 14673532 - 14773036 on NCBI Build GRCh38 | ||
Downstream | uncharacterized LOC152274 | LOC152274 | 122.7kb | Anti-sense | Chr. 3 - 14799331 - 14811414 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |