Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101928710 | LOC101928710 | -125.0kb | Sense | Chr. 17 - 78617384 - 78632057 on NCBI Build GRCh38 | |
Downstream | ubiquitin specific peptidase 36 | USP36 | DUB1 | 28.2kb | Sense | Chr. 17 - 78787381 - 78840898 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |