Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1958 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011518311.2 | XP_011516613.1 | 3 | 13898 | 92 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | chromosome 9 open reading frame 147 | C9orf147 | bA32M23.3 | -138.0kb | Anti-sense | Chr. 9 - 112409446 - 112490453 on NCBI Build GRCh38 |
Downstream | INTS3 and NABP interacting protein | INIP |
C9orf80, HSPC043, MISE, SOSSC, SSBIP1, hSSBIP1 |
55.9kb | Anti-sense | Chr. 9 - 112686506 - 112718107 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |