Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
RNA, U1 small nuclear 2 |
RNU1-2 |
RNU1C1, RNU1C2, U1C1, U1C21 |
-9.8kb | Anti-sense | Chr. 1 - 16895980 - 16896143 on NCBI Build GRCh38 |
Downstream | microfibrillar associated protein 2 | MFAP2 |
MAGP, MAGP-1, MAGP1 |
68.2kb | Sense | Chr. 1 - 16974502 - 16981649 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |