Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | insulin receptor substrate 2 | IRS2 | IRS-2 | -198.3kb | Sense | Chr. 13 - 109753837 - 109786567 on NCBI Build GRCh38 |
Downstream | long intergenic non-protein coding RNA 396 | LINC00396 | 66.7kb | Anti-sense | Chr. 13 - 110053285 - 110054952 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |