Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | transmembrane and tetratricopeptide repeat containing 2 | TMTC2 | IBDBP1 | Sense | Chr. 12 - 82686952 - 83134868 on NCBI Build GRCh38 | |
Downstream | solute carrier family 6 member 15 | SLC6A15 |
NTT73, SBAT1, V7-3, hv7-3 |
2.1mb | Anti-sense | Chr. 12 - 84859488 - 84912876 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |