Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RWD domain containing 4 | RWDD4 |
FAM28A, RWDD4A |
-43.4kb | Anti-sense | Chr. 4 - 183639635 - 183659219 on NCBI Build GRCh38 |
Downstream | storkhead box 2 | STOX2 | 91.8kb | Sense | Chr. 4 - 183796331 - 184023530 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |