Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | aryl-hydrocarbon receptor repressor | AHRR |
AHH, AHHR, bHLHe77 |
Anti-sense | Chr. 5 - 304176 - 438291 on NCBI Build GRCh38 | |
Downstream | EXOC3 antisense RNA 1 | EXOC3-AS1 | C5orf55 | 21.1kb | Sense | Chr. 5 - 441835 - 443160 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |