Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | nocturnin | NOCT |
CCR4L, CCRN4L, Ccr4c, NOC |
-102.7kb | Anti-sense | Chr. 4 - 139015759 - 139045939 on NCBI Build GRCh38 |
Downstream | mitochondria localized glutamic acid rich protein | MGARP |
C4orf49, CESP-1, HUMMR, OSAP |
114.6kb | Sense | Chr. 4 - 139266163 - 139280338 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |