Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | protein phosphatase 1 regulatory subunit 17 | PPP1R17 |
C7orf16, GSBS |
-146.8kb | Anti-sense | Chr. 7 - 31687017 - 31709931 on NCBI Build GRCh38 |
Downstream | phosphoribosyl pyrophosphate synthetase 2 pseudogene | LOC100130673 | 598.2kb | Sense | Chr. 7 - 32456897 - 32458494 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |