Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | small integral membrane protein 15 | SMIM15 | C5orf43 | Anti-sense | Chr. 5 - 61157709 - 61162475 on NCBI Build GRCh38 | |
Downstream | uncharacterized LOC101928630 | CTC-436P18.1 | 3.2kb | Sense | Chr. 5 - 61162316 - 61232080 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |