Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC284577 | LOC284577 | -49.6kb | Sense | Chr. 1 - 205456784 - 205469106 on NCBI Build GRCh38 | |
Downstream | uncharacterized LOC284578 | LOC284578 | 33.6kb | Anti-sense | Chr. 1 - 205554273 - 205556635 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |