Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006112.3 | NP_006103.1 | 10 | 108 | ||
NR_036543.1 | 10 | 108 | ||||
NR_036544.1 | 10 | 108 | ||||
GenBank mRNA | CK300365.1 | - | 1 | 232 | 108 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | hippocalcin like 4 | HPCAL4 | HLP4 | -61.6kb | Anti-sense | Chr. 1 - 39678648 - 39691710 on NCBI Build GRCh38 |
Downstream | bone morphogenetic protein 8b | BMP8B |
BMP8, OP2 |
411.0 | Anti-sense | Chr. 1 - 39757182 - 39788861 on NCBI Build GRCh38 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |